Skip to Content
Merck
Butyryl fentanyl solution 100 μg/mL in methanol, certified reference material, ampule of 0.5 mL, Cerilliant®

Butyryl fentanyl solution

Butyryl fentanyl solution 100 μg/mL in methanol, certified reference material, ampule of 0.5 mL, Cerilliant®
Empirical Formula (Hill Notation):
C23H30N2O
CAS Number:
Molecular Weight:
350.50

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing