
Brilliant green (hydrogen sulfate) (C.I. 42040)

Synonym(s):
Brilliant green (hydrogen sulfate) (C.I. 42040)
Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing