
Boc-Arg-OH

Synonym(s):
Nα-(tert-Butoxycarbonyl)-L-arginine, Nα-Boc-L-arginine
Linear Formula:
HN=C(NH2)NH(CH2)3CH(COOH)NHC(O)OC(CH3)3
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing