
Apraclonidine hydrochloride

Synonym(s):
p-Aminoclonidine monohydrochloride, 2-(4-Amino-2,6-dichloroanilino)-2-imidazoline hydrochloride
Empirical Formula (Hill Notation):
C9H10Cl2N4 · HCl
CAS Number:
Molecular Weight:
281.57
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2