Skip to Content
Merck
Alcohol Dehydrogenase, recombinant from E. coli ≥500 U/mL

Alcohol Dehydrogenase, recombinant from E. coli

Alcohol Dehydrogenase, recombinant from E. coli ≥500 U/mL
Synonym(s):
Alcohol:NADP+ oxidoreductase
CAS Number:

Products

Product Category
Brand
Color
Physical Form
Recombinant Host
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing