
Alcohol 100%

Synonym(s):
Alcohol
Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing