Skip to Content
Merck
Adenosine 5′-monophosphate sodium salt from yeast, ≥99%

Adenosine 5′-monophosphate sodium salt

Adenosine 5′-monophosphate sodium salt from yeast, ≥99%
Synonym(s):
5′-Adenylic acid, A-5′-P, AMP
Empirical Formula (Hill Notation):
C10H14N5O7P · xNa+ · yH2O
CAS Number:
Molecular Weight:
347.22 (anhydrous free acid basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing