Skip to Content
Merck
4-tert-Butylcalix[4]arene 95%

4-tert-Butylcalix[4]arene

4-tert-Butylcalix[4]arene 95%
Empirical Formula (Hill Notation):
C44H56O4
CAS Number:
Molecular Weight:
648.91

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing