Skip to Content
BS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4,4′-Methylenebis(
N
,
N
-dimethylaniline)
Synonym(s):
N
,
N
,
N
′,
N
′-Tetramethyl-4,4′-diaminodiphenylmethane,
N
,
N
,
N
′,
N
′-Tetramethyl-4,4′-methylenedianiline, Arnolds base, Michlers base
Linear Formula:
CH
2
[C
6
H
4
N(CH
3
)
2
]
2
CAS Number:
101-61-1
Molecular Weight:
254.37
EC Number:
202-959-2
Products
Product Category
dyes (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M44451
4,4′-Methylenebis(
N
,
N
-dimethylaniline),
97.5 - 102.5% purity, powder, crystals or chunks
Expand
View Pricing