Skip to Content
BS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Methyl-1-butanol
Synonym(s):
Isoamyl alcohol, Isopentyl alcohol, 3-Methyl-1-butanol
Linear Formula:
(CH
3
)
2
CHCH
2
CH
2
OH
CAS Number:
123-51-3
Molecular Weight:
88.15
EC Number:
204-633-5
Products
Product Category
solvents (5)
Brand
Sigma-Aldrich (5)
Supelco (2)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
ACS reagent (1)
Anhydrous (1)
analytical standard (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
M32658
3-Methyl-1-butanol,
reagent grade, 98%
Expand
View Pricing
320021
3-Methyl-1-butanol,
ACS reagent, ≥98.5%
Expand
View Pricing
309435
3-Methyl-1-butanol,
anhydrous, ≥99%
Expand
View Pricing
PHR2337
3-Methyl-1-butanol,
certified reference material, pharmaceutical secondary standard
Expand
View Pricing
1424153
3-Methyl-1-butanol,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
77664
3-Methyl-1-butanol,
analytical standard
Expand
View Pricing
15-1590
3-Methyl-1-butanol,
JIS special grade, ≥98.0%
Expand
View Pricing
15-1600
3-Methyl-1-butanol,
SAJ first grade, ≥96.0%
Expand
View Pricing