
3β-Taraxerol

Synonym(s):
(3β)-D-Friedoolean-14-en-3-ol, (3β13α)-13-Methyl-27-norolean-14-en-3-ol, Alnulin, Skimmiol, Taraxerol, Tiliadin
Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing