
2-Methylpyrazine

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique