Skip to Content
Merck
2-Methylallylamine hydrochloride

2-Methylallylamine hydrochloride

2-Methylallylamine hydrochloride
Synonym(s):
2-Methyl-2-propen-1-amine hydrochloride
Linear Formula:
H2C=C(CH3)CH2NH2 · HCl
CAS Number:
Molecular Weight:
107.58

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing