Skip to Content
Merck
2,4,5-Trichloropyrimidine 99%

2,4,5-Trichloropyrimidine

2,4,5-Trichloropyrimidine 99%
Empirical Formula (Hill Notation):
C4HCl3N2
CAS Number:
Molecular Weight:
183.42

Products

Product Category
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing