
1,4-Bis(dicyclohexylphosphino)butane

Linear Formula:
(C6H11)2P(CH2)4P(C6H11)2
CAS Number:
Molecular Weight:
450.66
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing