Skip to Content
Merck
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride 96%

1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride

1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride 96%
Empirical Formula (Hill Notation):
C10H11NO2 · HCl
CAS Number:
Molecular Weight:
213.66
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing