Skip to Content
Merck
All Photos(1)

Key Documents

V2881

Freezing vials, natural

capacity 500 μL, skirted bottom, non-sterile

Synonym(s):

cryopreservation tubes, cryopreservation vials, cryovials, freezing tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121704
NACRES:
NB.22
Pricing and availability is not currently available.

material

colorless tube
polypropylene tube
skirted bottom

sterility

non-sterile

feature

Internal thread

packaging

pack of 1000 ea

manufacturer/tradename

Simport T334-2

parameter

-196 °C min. temp.

technique(s)

cryopreservation: suitable

capacity

0.5 mL
500 μL

color

natural Cap

Looking for similar products? Visit Product Comparison Guide

General description

Manufactured from an exceptionally clear, virgin polypropylene with a medical-grade silicone O-ring seal. These vials are designed for safe storage at temperatures as low as -196°C but should be used only in the gas phase of liquid nitrogen.

The specially formulated silicone O-ring ensures a positive leakproof seal at all temperatures. A unique base on the skirted vials locks them into place in the microtube locking work station (Z376809). Tubes can be autoclaved to 121°C in upright position with caps loosened. They also can be centrifuged up to 17,000 × g.

Packaging

Caps are bagged separately from tubes
For non-sterile vials, caps and tubes are bagged separately.
For sterile vials, caps and tubes are assembled in bags.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What material is the freezing vial made from?

    1 answer
    1. The vials are made of polypropylene material.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service