Skip to Content
Merck
All Photos(1)

Key Documents

P9489

Sigma-Aldrich

Anti-p35 (Cdk5 Regulator) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CDK5P35, Anti-CDK5R, Anti-NCK5A, Anti-p23, Anti-p25, Anti-p35, Anti-p35nck5a

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

human, rat

technique(s)

microarray: suitable
western blot: 1:1,000 using a rat brain extract (cytosolic fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Cell division protein kinase 5 (Cdk5) is a serine/threonine kinase and shares homology to other Cdks. It belongs to cdk family.

Immunogen

synthetic peptide corresponding to the C-terminus of p35 (Cdk5R), (human, amino acids 192-209).

Application

Anti-p35 (Cdk5 Regulator) antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

Cell division protein kinase 5 (Cdk5) kinase activity can be detected mainly in postmitotic neurons in the central nervous system (CNS). Cdk5/p35 activity is required in the mature CNS. Cdk5 phosphorylates dopamine and cyclic adenosine 3′,5′-monophosphate-regulated phosphoprotein of 32 kDa (DARPP-32) and Syntaxin-binding protein 1 (Munc18), which in turn affects synaptic vesicle endocytosis. Deregulation of Cdk5 activity contributes to the pathogenesis of neurodegenerative diseases such as Alzheimer′s disease (AD). It mediates neuronal migration during cortical development. Cdk5-/- mice exhibit embryonic lethality associated with disruption of the cortical laminar structures in the cerebral cortex, olfactory bulb, hippocampus, and cerebellar cortex. Mice lacking p35 display defects in cortical lamination and fasciculation of axonal fibers. In developing mice brain, Cdk5/p35 interacts with Reelin/Dab1 and positions of cortical neurons.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of N-cadherin-mediated adhesion by the p35-Cdk5 kinase
Kwon YT, et al.
Current Biology, 10(7), 363-372 (2000)
Perinatal abrogation of Cdk5 expression in brain results in neuronal migration defects
Hirasawa M, et al.
Proceedings of the National Academy of Sciences of the USA, 101(16), 6249-6254 (2004)
Synergistic contributions of cyclin-dependant kinase 5/p35 and Reelin/Dab1 to the positioning of cortical neurons in the developing mouse brain
Ohshima T, et al.
Proceedings of the National Academy of Sciences of the USA, 98(5), 2764-2769 (2001)
Cyclin-dependent kinase 5/p35 contributes synergistically with Reelin/Dab1 to the positioning of facial branchiomotor and inferior olive neurons in the developing mouse hindbrain
Ohshima T, et al.
Neuroscience journal, 22(10), 4036-4044 (2002)
Activation of cyclin-dependent kinase 5 is a consequence of cell death
Ye Y, et al.
BioMed Research International, 2009 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service