Key Documents
EP6731010025
Eppendorf® CellXpert® C170i CO2 Incubator
Std, Left, AC/DC input 120 V, US 2-pin plug
Synonym(s):
Eppendorf CO2 Incubator
About This Item
UNSPSC Code:
41102404
feature
Std, Left
manufacturer/tradename
6731010025
Eppendorf®
parameter
4-50 °C temp. range
capacity
(170 L)
(740 L)
Related Categories
General description
Stay flexible for the future
  • Upgrade your device later in your lab (e.g. door handle position)
  • Advanced user interface for easy and complete control over parameter settings, monitoring, and documentation
Safety for your cells
  • Uniform temperature with multiple sensors verified at 27 points (German Norm DIN 12880)
  • Fast temperature and CO2 recovery in less than 5 minutes without overshoot
  • Vibration and turbulence protection with fan-less design
  • 180 ºC High Temperature Disinfection (HTD)
Save money
  • Significantly reduced gas consumption with smart gas control
  • Up to 25 % more usable space with fan-less design
  • No recurring costs for internal replacement parts (HEPA filters, UV lamps etc.)
Legal Information
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG