Skip to Content
Merck
All Photos(2)

Documents

AV44797

Sigma-Aldrich

Anti-RCE1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FACE2, Anti-RCE1 homolog, prenyl protein peptidase (S. cerevisiae), Anti-RCE1A, Anti-RCE1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

guinea pig, bovine, rat, rabbit, human, horse, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RCE1(9986)

Immunogen

Synthetic peptide directed towards the N terminal region of human RCE1

Application

Anti-RCE1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Ras converting CAAX endopeptidase 1 (RCE1) is a metalloproteinase required for the maintenance of CAAX-type prenylated proteins. The activity of RCE1 is essential for phosphodiesterase 6 trafficking and the survival of photoreceptor cells.

Sequence

Synthetic peptide located within the following region: WARCLTDMRWLRNQVIAPLTEELVFRACMLPMLAPCMGLGPAVFTCPLFF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey R Christiansen et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(21), 8862-8866 (2011-05-11)
Prenylation is the posttranslational modification of a carboxyl-terminal cysteine residue of proteins that terminate with a CAAX motif. Following prenylation, the last three amino acids are cleaved off by the endoprotease, RAS-converting enzyme 1 (RCE1), and the prenylcysteine residue is
J C Otto et al.
The Journal of biological chemistry, 274(13), 8379-8382 (1999-03-20)
Proteins containing C-terminal "CAAX" sequence motifs undergo three sequential post-translational processing steps: modification of the cysteine with either a 15-carbon farnesyl or 20-carbon geranylgeranyl isoprenyl lipid, proteolysis of the C-terminal -AAX tripeptide, and methylation of the carboxyl group of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service