Skip to Content
Merck
All Photos(1)

Key Documents

PHR3608

Supelco

N-Nitrosodibutylamine (NDBA)

certified reference material, pharmaceutical secondary standard

Synonym(s):

N-Butyl-N-nitroso-1-butanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O
CAS Number:
Molecular Weight:
158.24
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1466641

form

methanol solution

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

application(s)

pharmaceutical

SMILES string

N(N=O)(CCCC)CCCC

InChI

1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3

InChI key

YGJHZCLPZAZIHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

N-Nitrosodibutylamine (NDBA) pharmaceutical secondary reference standard is used for the determination of NDBA concentration in water samples via HPLC method in conjunction with a diode array detector.[1]

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Solvent

Description

    1000 ug/mL solution in methanol

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of a method for the simultaneous quantification of N-nitrosamines in water samples based on stir bar sorptive extraction combined with high-performance liquid chromatography and diode array detection
Talebpour, Z et al.
Journal of Separation Science, 38, 1601-1609 (2015)

Articles

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service