Skip to Content
Merck
All Photos(1)

Key Documents

91799

Supelco

Terbuthylazine-(ethyl-d5)

analytical standard

Synonym(s):

N2-tert-Butyl-6-chloro-N4-ethyl-d5-1,3,5-triazine-2,4-diamine, Terbuthylazine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D5H11ClN5
CAS Number:
Molecular Weight:
234.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

ClC1=NC(NC(C)(C)C)=NC(NC([2H])([2H])C([2H])([2H])[2H])=N1

InChI

1S/C9H16ClN5/c1-5-11-7-12-6(10)13-8(14-7)15-9(2,3)4/h5H2,1-4H3,(H2,11,12,13,14,15)/i1D3,5D2

InChI key

FZXISNSWEXTPMF-RPIBLTHZSA-N

General description

Terbuthylazine-(ethyl-d5)is an isotope-labeled analog of triazide herbicide terbuthylazine, widely used for vegetation management in forest plantations.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Persistence of terbuthylazine in soils
Sahid BI and Teoh SS
Bulletin of Environmental Contamination and Toxicology, 52(2), 226-230 (1994)
Sorption of the herbicide terbuthylazine in two New Zealand forest soils amended with biosolids and biochars
Wang H, et al.
Journal of Soils and Sediments, 10(2), 283-289 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service