Skip to Content
Merck
All Photos(1)

Key Documents

49739

Supelco

Glutathione reduced, immobilized on Agarose CL-4B

suitable for affinity chromatography, powder (lyophilized)

Synonym(s):

Reduced Glutathione Agarose Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.21

form

powder (lyophilized)

Quality Level

contains

50% lactose as stabilizer

concentration

~10 mM ligand: reduced glutathione (packed gel)

technique(s)

affinity chromatography: suitable

swelling

~70 mg swells to 1 mL gel

shipped in

wet ice

storage temp.

−20°C

General description

the lactose stabilizer must be removed prior to use by washing the gel onto filter with water or buffer; the reduced glutathione is attached through the sulfur to epoxy-activated 4% cross-linked beaded agarose (C10-spacer)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service