Skip to Content
Merck
All Photos(2)

Key Documents

32819

Supelco

Carazolol-d7

VETRANAL®, analytical standard

Synonym(s):

1-(Carbazol-4-yloxy)-3-(isopropyl-d7-amino)-2-propanol, 4-(2-Hydroxy-3-isopropyl-d7-amino-propoxy)carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D7H15N2O2
CAS Number:
Molecular Weight:
305.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+7

SMILES string

[2H]C([2H])([2H])C([2H])(NCC(O)COc1cccc2[nH]c3ccccc3c12)C([2H])([2H])[2H]

InChI

1S/C18H22N2O2/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16/h3-9,12-13,19-21H,10-11H2,1-2H3/i1D3,2D3,12D

InChI key

BQXQGZPYHWWCEB-QLWPOVNFSA-N

General description

Carazolol-d7 is an isotopically labeled compound of the β-blocker, carazolol.

Application

Carazolol-d7 may be used as an internal standard for the determination of carazolol in animal tissues and water samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and ultra-fast liquid chromatography-quadrupole/linear ion trap mass spectrometry (UFLC-Q/TRAP MS), respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of nine beta-blockers in porcine tissues by ultra-fast liquid chromatography coupled with quadrupole/linear ion trap mass spectrometry.
Zhang H, et al.
Se Pu / Chinese Journal of Chromatography, 32(6), 573-581 (2014)
A Kaufmann et al.
Rapid communications in mass spectrometry : RCM, 15(18), 1747-1751 (2001-09-14)
A fast and simple method for the quantification of a number of tranquilizers and the beta-blocker Carazolol in pork and bovine kidney is described. Extracts are purified/concentrated by a solid phase extraction step and separated on a reversed phase column
Validating the rejection of trace organic chemicals by reverse osmosis membranes using a pilot-scale system.
Fujioka T, et al.
Desalination, 358, 18-26 (2015)
Jian-Qiao Cheng et al.
Molecules (Basel, Switzerland), 24(4) (2019-03-03)
This paper presents an application of high performance liquid chromatography coupled with quadrupole orbitrap high-resolution mass spectrometry (HPLC-Q-Orbitrap HRMS) for the analysis of 27 β-blockers and metabolites in milk powder. Homogenized milk power samples were extracted by acetonitrile and purified
Lenise Guimarães de Oliveira et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(1), 32-39 (2016-11-22)
A method for the determination of five sedatives and 14 β-blocker residues in swine, bovine and equine kidney was validated. Samples were extracted with acetonitrile and purified using dispersive solid phase extraction (d-SPE) with Celite 545 with subsequent analysis by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service