Skip to Content
Merck
All Photos(1)

Key Documents

03-1440

Sigma-Aldrich

Benzoyl chloride

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCl
CAS Number:
Molecular Weight:
140.57
Beilstein:
471389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.88 (vs air)

vapor pressure

1 mmHg ( 32 °C)

Assay

≥98.0%

autoignition temp.

1056 °F

expl. lim.

4.9 %

availability

available only in Japan

refractive index

n20/D 1.553 (lit.)

bp

198 °C (lit.)

mp

−1 °C (lit.)

density

1.211 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccccc1

InChI

1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

PASDCCFISLVPSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong Li et al.
Journal of the American Society for Mass Spectrometry, 24(4), 493-501 (2013-03-21)
The benzoxyl radical is a key intermediate in the combustion of toluene and other aromatic hydrocarbons, yet relatively little experimental work has been performed on this species. Here, a combination of electrospray ionization (ESI), multistage mass spectrometry experiments, and density
Russell J Cox et al.
Chemical communications (Cambridge, England), (8)(8), 1037-1039 (2005-02-19)
Conditions are reported for the facile, high-yielding coupling of acyl chlorides with terminal alkynes in a reaction involving palladium and copper iodide; the reaction is tolerant of a wide variety of acyl chlorides and terminal alkynes and provides a convenient
Sarah D Lever et al.
Journal of hazardous materials, 115(1-3), 91-100 (2004-11-03)
The risks associated with batch processing in the manufacture of chemicals and pharmaceuticals via highly exothermic reactions are of special interest due to the possibility of runaway reactions. o-Nitrated benzoyl chlorides are intermediates in the production of agrochemicals and are
Pramod Kumar Verma et al.
Physical chemistry chemical physics : PCCP, 11(38), 8467-8476 (2009-09-24)
In this contribution we attempt to correlate the dynamical states of water molecules in reverse micelles with a solvolysis reaction in accordance with the activation energy barrier crossing model at the micellar interface. Precise measurement of the different dynamical states
Ran Liu et al.
Journal of mass spectrometry : JMS, 47(10), 1341-1346 (2012-09-29)
A high-performance liquid chromatography coupled with Q-time of flight mass spectrometry (HPLC/Q-TOF MS) method was developed and validated for the determination of 1, 3-diaminopropane, putrescine, cadaverine, spermidine and spermine in human plasma. The plasma samples were first pretreated by 10%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service