Skip to Content
Merck
All Photos(8)

Key Documents

RDYFCON05

Millipore

Female Connectors for ReadyStream® Bag

Adapters for ReadyStream® Bag – Tube connection

Synonym(s):

Tube Connector, Tube adaptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102014
NACRES:
NB.51

material

PEEK

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 5 connector

application(s)

food and beverages

General description

The autoclavable ReadyStream® female bag connector allows connection of the ReadyStream® bag outlet to tubes of 6.4 mm diameter (e.g. RDYTUB08). In that way the ReadyStream® system can be connected to a standard GL45 lab bottle where the media can be filled in. From this bottle, serving as media reservoir, a dilutor connected to this bottle can pull the media and dispense it to the sample.
For clean operations, caps for each connector are contained in this reference as well, so that the connectors can be sealed if needed.

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service