Skip to Content
Merck
All Photos(1)

Key Documents

PX0170

Supelco

Pentane

Synonym(s):

acs solvents

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-692-4

vapor density

2.48 (vs air)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

impurities

≤0.01% Water (H2O)

evapn. residue

≤0.001%

color

APHA: ≤30

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

density

~0.63 g/cm3 at 20 °C
0.626 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Evaluation of selected solid adsorbents for passive sampling of atmospheric oil and natural gas non-methane hydrocarbons.: This study evaluates solid adsorbents for sampling atmospheric hydrocarbons, including pentane, providing insights into environmental monitoring techniques. (Helmig et al., 2022).

  • Online Liquid Chromatography-Raman Spectroscopy Using the Vertical Flow Method.: This study introduces an online method combining liquid chromatography and Raman spectroscopy for analyzing compounds like pentane, enhancing analytical capabilities in complex matrices. (Lo and Hiramatsu, 2020).

  • An eNose-based method performing drift correction for online VOC detection under dry and humid conditions.: This paper presents an electronic nose method with drift correction for detecting volatile organic compounds, including pentane, under varying environmental conditions. (Jaeschke et al., 2020).

Analysis Note

Assay (GC): 98% min
Color (APHA): 30 max
Evaporation residue: 0.001% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Water (H2O): 0.01% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service