Skip to Content
Merck
All Photos(1)

Key Documents

8.41089

Sigma-Aldrich

2-Methyl-3-buten-2-ol

for synthesis

Synonym(s):

2-Methyl-3-buten-2-ol, Dimethyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-068-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

137 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

380 °C

potency

1800 mg/kg LD50, oral (Rat)

expl. lim.

1.5-9.4 % (v/v)

pH

9-10 (20 °C, 100 g/L in H2O)

bp

98-99 °C/1013 hPa

mp

-28 °C

transition temp

flash point 11 °C

density

0.824 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(C)(C)C=C

InChI

1S/C5H10O/c1-4-5(2,3)6/h4,6H,1H2,2-3H3

InChI key

HNVRRHSXBLFLIG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.822 - 0.826
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service