Skip to Content
Merck
All Photos(2)

Key Documents

79129BC

Millipore

GDS PPMX Riser

BioControl, For use with GDS PickPen® PIPETMAX®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121521

description

For use with GDS PICKPEN™ PIPETMAX™ System

manufacturer/tradename

BioControl Systems 79129

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

storage temp.

room temp

Components

One PPMX Riser

Compatibility

BioControl GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service