Skip to Content
Merck
All Photos(1)

Key Documents

71115

Sigma-Aldrich

RiboJuice siRNA Transfection Reagent

Efficient delivery of siRNA for targeted gene suppression

Synonym(s):

siRNA Transfection Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106502
NACRES:
NA.55

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

do not freeze

technique(s)

transfection: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

RNA interference experiments using siRNArequire a transfection reagent specificallyoptimized for siRNA delivery. RiboJuicesiRNA Transfection Reagent efficiently deliverssiRNA into a wide range of mammaliancell lines for targeted gene suppression.RiboJuice is an optimized, easy-to-use siRNAtransfection reagent formulated for minimaltoxicity. In addition, RiboJuice is compatiblewith GeneJuice Transfection Reagent forcotransfection of siRNA and plasmid DNAfor normalizing transfection efficiency with areporter gene.

RiboJuice Transfection Reagent has beensuccessfully used for gene suppression in avariety of cell lines.

Warning

Toxicity: Flammable (J)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Elbashir, S.M., et al. 2001. Nature411, 494.

Legal Information

RiboJuice is a trademark of EMD Biosciences, Inc., Madison, WI USA
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eeva Kaisa Rajakylä et al.
Cell reports, 30(12), 4266-4280 (2020-03-27)
Defects in the maintenance of intercellular junctions are associated with loss of epithelial barrier function and consequent pathological conditions, including invasive cancers. Epithelial integrity is dependent on actomyosin bundles at adherens junctions, but the origin of these junctional bundles is
Deivid C Rodrigues et al.
Cell reports, 30(12), 4179-4196 (2020-03-27)
Regulation of translation during human development is poorly understood, and its dysregulation is associated with Rett syndrome (RTT). To discover shifts in mRNA ribosomal engagement (RE) during human neurodevelopment, we use parallel translating ribosome affinity purification sequencing (TRAP-seq) and RNA
Reza Nadafi et al.
Cell reports, 30(12), 4110-4123 (2020-03-27)
Within lymph nodes (LNs), T follicular helper (TFH) cells help B cells to produce antibodies, which can either be protective or autoreactive. Here, we demonstrate that murine LN stromal cells (LNSCs) suppress the formation of autoreactive TFH cells in an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service