Skip to Content
Merck
All Photos(1)

Key Documents

5.00582

Sigma-Aldrich

Phorbol-12-myristate-13-acetate

≥98% (HPLC), liquid, PKC activator, Calbiochem®

Synonym(s):

InSolution Phorbol-12-myristate-13-acetate, 12-O-Tetradecanoyl-phorbol-13-acetate, PMA, TPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H56O8
CAS Number:
Molecular Weight:
616.83
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Phorbol-12-myristate-13-acetate, InSolution, ≥98%, 10 mM in DMSO, extremely potent mouse skin tumor promoter

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

O([C@]21[C@H]([C@H]3[C@@]([C@H]4[C@](CC(=C3)CO)(C(=O)C(=C4)C)O)([C@@H]([C@H]2OC(=O)CCCCCCCCCCCCC)C)O)C1(C)C)C(=O)C

InChI

1S/C36H56O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,24,27-28,30,32,37,41-42H,7-18,21-22H2,1-6H3/t24-,27+,28-,30-,32-,34-,35-,36-/m1/s1

InChI key

PHEDXBVPIONUQT-RGYGYFBISA-N

General description

Most commonly-used phorbol ester. Extremely potent mouse skin tumor promoter. Activates protein kinase C in vivo and in vitro, even at nM concentrations. Promotes the expression of iNOS in cultured hepatocytes. Activates Ca2+-ATPase and potentiates forskolin-induced cAMP formation. Inhibits apoptosis induced by the Fas antigen, but induces apoptosis in HL-60 promyelocytic leukemia cells. Its binding is reversible.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Reversible: yes
Secondary Target
Ca2+-ATPase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Use only fresh DMSO for preparing further dilutions.

Other Notes

Powell, D.J., et al. 2003. Mol. Cell Biol.23, 7794.
Tepper, C.G., et al. 1995. Proc. Natl. Acad. Sci. USA92, 8443.
Oishi, K., and Yamaguchi, M. 1994. J. Cell. Biochem.55, 168.
Macfarlane, D.E., and O′Donnell, P.S. 1993. Leukemia7, 1846.
Ghandi, V.C. and Jones, D.J., 1992. Neuropharmacol.31, 1101.
Hortelano, S., et al. 1992. J. Biol. Chem.267, 24937.
Kontny, E., et al. 1992. Eur. J. Pharmacol.227, 333.
Zanaboni, P.B., et al. 1992. J. Appl. Physiol.73, 2011.
Saltis, J., et al. 1991. J. Biol. Chem.266, 261.
Beguinot, L., et al. 1985. Proc. Natl. Acad. Sci. USA82, 2774.
Perchellet, J. 1985. Carcinogenesis6, 567.
Nishizuka, Y. 1984. Science255, 1365.
Mastro, A. 1982. Lymphokines6, 263.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

188.6 °F - (Dimethylsulfoxide)

Flash Point(C)

87 °C - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This study presents an example of a typical ELISpot assay for quantifying the number of pre-existing, antigen-reactive T cells from peripheral blood mononuclear cell (PBMC) samples obtained from donors previously infected with cytomegalovirus (CMV).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service