124036-M
Akt Inhibitor XVIII, SC66
The Akt Inhibitor XVIII, SC66 controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Akt Inhibitor XVIII, SC66, (2 E,6 E)-2,6- bis(4-Pyridylmethylene)cyclohexanone, (2E,6E)-2,6-bis(4-Pyridylmethylene)cyclohexanone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pale yellow
solubility
DMSO: 25 mg/mL
storage temp.
2-8°C
General description
A cell-permeable pyridine compound that represses Akt activation by allosterically disrupting Akt-PH domain binding to PIP3 and directly facilitating Akt ubiquitination with little proteasomal or deubiquitination activity. Blocks phosphorylations of Akt-Ser473 and downstream targets TSC, FOXO and PRAS40 in HEK293T cells at 8 µg/ml with minimal effect on pAkt-Thr450. Shown to be highly efficacious towards PI 3-K inhibitor resistant Akt1-E17K mutant and exert synergistic apoptotic activity with LY 294002 (Cat. No. 440202) against HeLa cells. Suppresses tumor growth in a mouse HEK293T-xenograft model (30 mg/kg, s.c., twice per week).
A cell-permeable pyridine compound that selectively targets Akt, but not PLCδ, PH domain, preventing Akt membrane localization/activation and rendering Akt in a conformation susceptible to ubiquitination and subsequent proteasomal degradation. Akti-1/2 (Cat. Nos. 124017 & 124018) competes for Akt PH domain binding and blocks SC66-induced Akt ubiquitination and proteasomal degradation. Effectively inhibits HEK293T growth both in vitro (>99 inhibition at 0.4 M) and in mice in vivo (84% of control tumor size with twice weekly i.p. dose of 30 mg/kg), Synergizes with LY294002 (Cat. Nos. 440202 & 440204) in cell death induction in HeLa and HS-Sultan cultures in vitro.
Warning
Toxicity: Regulatory Review (Z)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Other Notes
Jo, H., et al. 2011. Proc. Natl. Acad. Sci. USA108, 6486.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service