Skip to Content
Merck
All Photos(1)

Key Documents

1.15127

Supelco

Ion exchanger Amberlite MB-3

mixed bed ion exchanger with colour indicator, strongly acidic H⁺ form and strongly basic OH⁻ form

Synonym(s):

Ion exchanger Amberlite MB-3

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41115711

form

resin

technique(s)

HPLC: suitable
LPLC: suitable

matrix

PS-DVB

particle size

300-1180 μm

density

0.69 g/cm3 at 20 °C

bulk density

700 kg/m3

separation technique

mixed mode

Analysis Note

Description: blue and cream-coloured spherules
Anion exchanger capacity: ≥ 0.6 mol/l
Kation exchanger capacity: ≥ 0.5 mol/l
Particle size (0.3 - 1.18 mm): ≥ 80 %

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service