Skip to Content
Merck
All Photos(1)

Key Documents

O-031

Supelco

Oxymorphone-D3-3-β-D-glucuronide solution

100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Oxymorphone Glucuronide Metabolite Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23D3H24NO10
Molecular Weight:
480.48
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])N1CC[C@]23[C@H]4Oc5c(OC6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)ccc(C[C@@H]1[C@]2(O)CCC4=O)c35

InChI

1S/C23H27NO10/c1-24-7-6-22-13-9-2-3-11(32-21-16(28)14(26)15(27)18(34-21)20(29)30)17(13)33-19(22)10(25)4-5-23(22,31)12(24)8-9/h2-3,12,14-16,18-19,21,26-28,31H,4-8H2,1H3,(H,29,30)/t12-,14+,15+,16-,18+,19+,21?,22+,23-/m1/s1/i1D3

InChI key

ILRLBQVIWURYLX-AKWUMPEOSA-N

General description

A stable-labeled internal standard suitable for quantitation of oxymorphone levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, pain prescription monitoring, clinical toxicology, forensic analysis, pharmaceutical research, or isotope dilution methods. Oxymorphone-3ß-D-glucuronide is a major metabolite of oxymorphone in urine and plasma. Oxymorphone, sold under trade names such as Opana®, Numorphan®, and Numorphone, is an opiate analgesic indicated for the relief of moderate to severe pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Numorphan is a registered trademark of Endo Pharmaceuticals Inc.
Opana is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service