Key Documents
Z214388
Ace-Thred Thermometer/Tubing Adapter
With hose connection, joint: ST/NS 29/42, thread I.D. 9.0-10.5 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
label
F
Joint
joint: ST/NS29/42
manufacturer/tradename
Ace Glass 526140
thread I.D.
9.0-10.5 mm
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)