Skip to Content
Merck
All Photos(1)

Key Documents

W438700

Sigma-Aldrich

2-Pentylthiophene

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14S
CAS Number:
Molecular Weight:
154.27
FEMA Number:
4387
MDL number:
UNSPSC Code:
12164502
Flavis number:
15.096
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

Assay

≥98%

composition

May contain triglycerol and tert-Butyl methyl ether

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; meaty

InChI

1S/C9H14S/c1-2-3-4-6-9-7-5-8-10-9/h5,7-8H,2-4,6H2,1H3

InChI key

NOYVOSGVFSEKPR-UHFFFAOYSA-N

Legal Information

For R&D use and/or Food Manufacture Only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F

Flash Point(C)

75 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service