SYNA972440
Synthware™ distilling adapter with compression cap
joint: ST/NS 24/40, side arm angle: 75 deg
Synonym(s):
Synthware A972440, glass distillation adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A972440
General description
With compression cap (RC009) and Viton® O-ring for forming a vacuum tight seal of plain thermometer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company