Skip to Content
Merck
All Photos(1)

Key Documents

B30405

Sigma-Aldrich

4-Benzylpyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N
CAS Number:
Molecular Weight:
169.22
Beilstein:
115831
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.582 (lit.)

bp

287 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

SMILES string

C(c1ccccc1)c2ccncc2

InChI

1S/C12H11N/c1-2-4-11(5-3-1)10-12-6-8-13-9-7-12/h1-9H,10H2

InChI key

DBOLXXRVIFGDTI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niina Tani et al.
European journal of medicinal chemistry, 47(1), 270-277 (2011-11-22)
Because of the increasing number of immunocompromised patients and due to problems with antifungal treatment, especially with the most widely used antifungals, azoles, there is an urgent need for new, potent and safe antifungals with fewer cytochrome P450 (CYP)-mediated interactions
Manish B Shah et al.
Molecular pharmacology, 80(6), 1047-1055 (2011-08-31)
The biochemical, biophysical, and structural analysis of the cytochrome P450 2B subfamily of enzymes has provided a wealth of information regarding conformational plasticity and substrate recognition. The recent X-ray crystal structure of the drug-metabolizing P450 2B6 in complex with 4-(4-chlorophenyl)imidazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service