Skip to Content
Merck
All Photos(1)

Key Documents

375608

Sigma-Aldrich

2-Bromopropane-d7

98 atom % D

Synonym(s):

Isopropyl-d7 bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2CDBr
CAS Number:
Molecular Weight:
130.03
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

59 °C (lit.)

mp

-89 °C (lit.)

density

1.383 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H]C([2H])([2H])C([2H])(Br)C([2H])([2H])[2H]

InChI

1S/C3H7Br/c1-3(2)4/h3H,1-2H3/i1D3,2D3,3D

InChI key

NAMYKGVDVNBCFQ-YYWVXINBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Repr. 1A - STOT RE 2 Inhalation

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service