Skip to Content
Merck
All Photos(1)

Key Documents

365009

Sigma-Aldrich

2-Bromo-5-fluorotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Br)F
CAS Number:
Molecular Weight:
189.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

177 °C/756 mmHg (lit.)

density

1.495 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(F)ccc1Br

InChI

1S/C7H6BrF/c1-5-4-6(9)2-3-7(5)8/h2-4H,1H3

InChI key

RJPNVPITBYXBNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Bromo-5-Fluorobenzoic Acid.
Junhong ZYL.
Guangzhou Chemical Industry/ Guangzhou Huagong, 12, 2253-2255 (2000)
A Concise and Efficient Synthesis of [2-Methyl-5-methylsulfonyl-4-(pyrrol-1-yl) benzoyl] guanidinium Methanesulfonate (Eniporide).
Baumgarth M and Gericke R.
European Journal of Organic Chemistry, 12, 2253-2255 (2000)
Peri fluoro steric effects: syntheses and comparative acid-catalyzed isomerization of the 8-, 9-, and 11-fluoro-1, 2, 3, 4-tetrahydro-7, 12-dimethylbenz [a] anthracenes to exo methylene tautomers.
Witiak DT, et al.
The Journal of Organic Chemistry, 53(2), 345-352 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service