Merck
Methyltransferase-like protein 16 binds the 3'-terminal triple helix of MALAT1 long noncoding RNA.
Proceedings of the National Academy of Sciences of the United States of America (2016-11-23)
Jessica A Brown, Charles G Kinzig, Suzanne J DeGregorio, Joan A Steitz
RESUMO
Metastasis-associated lung adenocarcinoma transcript 1 (MALAT1), a cancer-promoting long noncoding RNA, accumulates in cells by using a 3'-triple-helical RNA stability element for nuclear expression (ENE). The ENE, a stem-loop structure containing a U-rich internal loop, interacts with a downstream A-rich tract (ENE+A) to form a blunt-ended triple helix composed of nine U•A-U triples interrupted by a C•G-C triple and C-G doublet. This unique structure prompted us to explore the possibility of protein binding. Native gel-shift assays revealed a shift in radiolabeled MALAT1 ENE+A RNA upon addition of HEK293T cell lysate. Competitive gel-shift assays suggested that protein binding depends not only on the triple-helical structure but also its nucleotide composition. Selection from the lysate using a biotinylated-RNA probe followed by mass spectrometry identified methyltransferase-like protein 16 (METTL16), a putative RNA methyltransferase, as an interacting protein of the MALAT1 ENE+A. Gel-shift assays confirmed the METTL16-MALAT1 ENE+A interaction in vitro: Binding was observed with recombinant METTL16, but diminished in lysate depleted of METTL16, and a supershift was detected after adding anti-METTL16 antibody. Importantly, RNA immunoprecipitation after in vivo UV cross-linking and an in situ proximity ligation assay for RNA-protein interactions confirmed an association between METTL16 and MALAT1 in cells. METTL16 is an abundant (∼5 × 10
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-METTL16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution