Merck
  • An unsymmetric cisplatin-based Pt(iv) derivative containing 2-(2-propynyl)octanoate: a very efficient multi-action antitumor prodrug candidate.
An unsymmetric cisplatin-based Pt(iv) derivative containing 2-(2-propynyl)octanoate: a very efficient multi-action antitumor prodrug candidate.
Dalton transactions (Cambridge, England : 2003) (2017-10-07)
Elisabetta Gabano, Mauro Ravera, Ilaria Zanellato, Stefano Tinello, Andrea Gallina, Beatrice Rangone, Valentina Gandin, Cristina Marzano, Maria Grazia Bottone, Domenico Osella
RESUMO
The design, synthesis, characterization and biological properties of a Pt(iv) complex containing the very active inhibitor of histone deacetylase (2-propynyl)octanoic acid, POA, as an axial ligand are reported here. The title complex, namely (OC-6-44)-acetatodiamminedichlorido(2-(2-propynyl)octanoato)platinum(iv), 1, containing POA in racemic or in enantiomeric forms, was one/two orders of magnitude more active than cisplatin, depending on the chemo-sensitivity of the cancer cell lines. Moreover, 1 exhibited similar or even better antiproliferative activity than (OC-6-33)-diamminedichloridobis(2-propylpentanoato)platinum(iv), 2, containing two molecules of the well-known histone deacetylase inhibitor 2-propylpentanoic (valproic) acid. The high potency of 1 is likely due to its high cellular accumulation and to the synergism between the DNA-damaging cisplatin and the histone deacetylase inhibitor POA, both released upon the intracellular reduction of 1. Prodrug 1, after oral administration, caused an impressive reduction of the tumor mass (94%) in a model of solid tumor (murine Lewis lung carcinoma), compared to that of the control, whereas (intraperitoneal) cisplatin induced a tumor regression of 75% only. A good accumulation of 1 was observed in the tumor mass. The time course of the body weight attested that cisplatin induced elevated anorexia, whereas treatment with 1 did not induce significant body weight loss throughout the therapeutic experiment.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder