Merck
  • Φ-order spectrophotokinetic characterisation and quantification of trans-cis oxyresveratrol reactivity, photodegradation and actinometry.
Φ-order spectrophotokinetic characterisation and quantification of trans-cis oxyresveratrol reactivity, photodegradation and actinometry.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-07-12)
Mounir Maafi, Mohammed Ahmed Al-Qarni
RESUMO
A new Φ-order kinetic method was proposed in this study for the investigation of trans-cis photoisomerization reaction of Oxyresveratrol (ORVT) subjected to non-isosbestic irradiation. In ethanolic media, it has been proven that forward (Φ
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Oxyresveratrol, ≥97.0% (HPLC)