Merck
  • Increasing dietary sugar concentration may improve dry matter intake, ruminal fermentation, and productivity of dairy cows in the postpartum phase of the transition period.
Increasing dietary sugar concentration may improve dry matter intake, ruminal fermentation, and productivity of dairy cows in the postpartum phase of the transition period.
Journal of dairy science (2009-06-17)
G B Penner, M Oba
RESUMO
The current study was undertaken to investigate the effect of feeding diets varying in sugar concentration to postpartum transition cows on productivity, ruminal fermentation, and nutrient digestibility. We hypothesized that the high-sugar diet would increase dry matter intake and lactation performance. The secondary objective was to characterize changes in ruminal fermentation and nutrient digestibility over the first 4 wk of lactation. Fifty-two Holstein cows, including 28 primiparous and 24 multiparous cows, 10 of which were previously fitted with a ruminal cannula, were assigned to the experimental diets containing either high sugar (HS = 8.4%) or low sugar (LS = 4.7%) immediately after calving, based on their expected calving date. Data and samples were collected on d 5.2 +/- 0.3, 12.2 +/- 0.3, 19.2 +/- 0.3, and 26.1 +/- 0.3 relative to parturition for wk 1, 2, 3, and 4 respectively. Cows fed HS had increased dry matter intake compared with those fed LS (18.3. vs. 17.2 kg/d). Further, cows fed HS sorted for particles retained on the pan of the Penn State Particle Size Separator to a greater extent than cows fed LS. Feeding HS tended to increase nadir (5.62 vs. 5.42), mean (6.21 vs. 6.06), and maximum pH (6.83 vs. 6.65). The duration (h/d) and area (pH x min/d) that ruminal pH was below pH 5.8 were not affected by treatment. Ruminal volatile fatty acid concentration and molar proportions of individual volatile fatty acids were not affected by treatment. The digestibility of dry matter, organic matter, neutral detergent fiber, and starch were not affected by treatment, averaging 63.3, 65.2, 43.2, and 93.5%, respectively. Feeding HS decreased plasma glucose concentration compared with feeding LS (51.3 vs. 54.0 mg/dL), but concentration of plasma insulin was not affected by treatment, averaging 4.17 microIU/mL. Cows fed HS had higher concentrations of plasma beta-hydroxybutrate (17.5 vs. 10.5 mg/dL) and nonesterified fatty acids (344 vs. 280 microEq/L). Milk yield and milk composition were not affected by treatment, but a tendency for increased milk fat yield was observed for cows fed HS compared with LS (1.44 vs. 1.35 kg/d). The results of the current study imply that replacing cracked corn grain with sucrose may improve dry matter intake, ruminal pH status, and lactation performance.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
PGO Enzyme Preparation, 1 G capsules
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination