Merck
  • Blood glucose kinetics and physiological changes in a type 1 diabetic finisher of the Ultraman triathlon: a case study.
Blood glucose kinetics and physiological changes in a type 1 diabetic finisher of the Ultraman triathlon: a case study.
European journal of applied physiology (2017-03-18)
Christopher W Bach, Daniel A Baur, William S Hyder, Michael J Ormsbee
RESUMO
To investigate the blood glucose kinetics and physiological effects experienced by a type 1 diabetic (T1D) finisher of a 3-day, multi-stage ultra endurance triathlon consisting of a 10 km swim and 144.8 km bike (stage 1), a 275.4 km bike (stage 2), and an 84.4 km run (stage 3). The athlete self-monitored blood glucose (SMBG) levels via fingerstick blood draw and hand-held glucometer. Researchers evaluated blood glucose kinetics via a continuous glucose monitoring device. The athlete maintained normal dietary and insulin patterns before, during and after competition daily. Weight and body composition were measured via bioelectrical impedance and select biomarkers were measured in blood. The athlete spent 73.0, 3.4, and 15.1% of during race time in a hyperglycemic state (≥130 mg dL It is possible for a T1D athlete to complete a multi-stage ultraendurance triathlon and maintain glycemic control using SMBG methods. In addition, a T1D athlete participating in an ultraendurance triathlon results in substantial changes in body composition, hormones, and muscle damage.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Creatine Kinase Activity Assay Kit, sufficient for 100 tests (Ultraviolet)