Merck
FADD regulates NF-κB activation and promotes ubiquitination of cFLIPL to induce apoptosis.
Scientific reports (2016-03-15)
Kishu Ranjan, Chandramani Pathak
RESUMO
Tumor Necrosis Factor-α canonically induces the activation of NF-κB and associated gene product cellular FLICE-like inhibitory protein (cFLIPL) to promote cell survival. Previously, we demonstrated that ectopic expression of the Fas associated death domain (FADD) diminishes the expression of cFLIPL and transduces caspases-8 mediated apoptosis, independent of FasL stimulation in HEK 293T cells. However, the underlying molecular mechanism of FADD mediated ablation of cFLIP and NF-κB signaling to determining the fate of cell death or survival remains elusive. Here, we explored a novel molecular mechanism of FADD mediated apoptotic cell death that was directed by ubiquitination of cFLIPL and inhibition of NF-κB activation, independent of TNF-α stimulation. We found that induced expression of FADD firmly interacts with procaspase-8 and precludes cFLIPL to from the death inducing signaling complex (DISC). In addition, FADD negatively regulates cellular inhibitor of apoptosis protein 2 (cIAP2) and Bcl-2. Furthermore, FADD restrains cIAP2 expression and interacts with RIP1 and procaspase-8 to accomplish apoptotic cell death signaling. Interestingly, FADD was also found to promote JNK1 mediated activation of E3 ubiquitin ligase ITCH to degrade cFLIPL that may lead to commencement of apoptosis. Thus, FADD is an important regulator for determining the fate of cell death or survival.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-ITCH antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Tamanho da embalagem
Disponibilidade
Preço
Quantidade