Merck
Cell-length-dependent microtubule accumulation during polarization.
Current biology : CB (2010-05-25)
Dominique Seetapun, David J Odde
RESUMO
Breaking cell symmetry, known as polarization, requires dynamic reorganization of microtubules (MTs) and is essential to many cellular processes, including axon formation in neurons. A critical step in polarization is believed to be the "selective stabilization" of MTs, which hypothesizes a spatial and/or temporal shift toward net MT assembly in a preferred direction of growth. We now find that a simpler "length-dependent" model, in which MT assembly parameters are spatially and temporally constant, predicts MT accumulation in the direction of growth because of longer mean first passage times in the longer direction. We experimentally tested both models by tracking MT assembly dynamics in polarizing embryonic chick forebrain neurons, and we confirmed that assembly is spatially and temporally constant during axon formation. Cell polarization occurs most simply through cell-length-dependent accumulation of MTs without MT stabilization or capture. In this way, F-actin-mediated cell shape and size changes can be read out by dynamic MTs undergoing simple dynamic instability to ultimately break cell symmetry.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-EB1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution