Merck
CD19 monoclonal antibody HD37 inhibits anti-immunoglobulin-induced B cell activation and proliferation.
Journal of immunology (Baltimore, Md. : 1950) (1987-05-01)
A Pezzutto, B Dörken, P S Rabinovitch, J A Ledbetter, G Moldenhauer, E A Clark
RESUMO
The 95 Kd CD19 antigen is the broadest lineage specific surface marker for B cells: it is present on the surface of virtually all B lymphocytes, including early B progenitor cells. In this study we have evaluated the function of the CD19 antigen by using the CD19 mAb HD37. Binding of HD37 mAb to B cells at low doses (0.5 microgram/ml) induced a strong inhibition of the proliferative response to anti-Ig. This inhibition was not mediated by the Fc portion of the antibody, since F(ab')2 fragments were as effective as the whole antibody. Both dose-response curve analysis and experiments in which a cross-linking second step anti-mouse antibody was added suggested that cross-linking of CD19 antigens was necessary for optimal inhibition. Early phases in B cell activation were affected by the HD37 mAb: it significantly reduced the number of cells that left G0 and entered the G1 phase of the cell cycle upon triggering with anti-mu. The increase in free intracellular ionized calcium [Ca2+]i that is induced by anti-mu was also consistently reduced by CD19 mAb. Cross-linking was also crucial for this effect, suggesting that a causal relationship may exist between the inhibition of anti-Ig-mediated [Ca2+]i fluxes and inhibition of proliferation. A variable but clear increase in [Ca2+]i levels followed cross-linking of CD19 antigens by specific mAb. This evidence suggests that CD19 molecules may function in the downregulation of B cell growth and proliferation.