Merck
  • Molecular Characterization of Carbapenem-Nonsusceptible Enterobacterial Isolates Collected during a Prospective Interregional Survey in France and Susceptibility to the Novel Ceftazidime-Avibactam and Aztreonam-Avibactam Combinations.
Molecular Characterization of Carbapenem-Nonsusceptible Enterobacterial Isolates Collected during a Prospective Interregional Survey in France and Susceptibility to the Novel Ceftazidime-Avibactam and Aztreonam-Avibactam Combinations.
Antimicrobial agents and chemotherapy (2015-10-21)
Hervé Dupont, Olivier Gaillot, Anne-Sophie Goetgheluck, Claire Plassart, Jean-Philippe Emond, Marion Lecuru, Nicolas Gaillard, Sarah Derdouri, Baptiste Lemaire, Marion Girard de Courtilles, Vincent Cattoir, Hedi Mammeri
RESUMO
An interregional surveillance program was conducted in the northwestern part of France to determine the prevalence of carbapenem-nonsusceptible Enterobacteriaceae (CNSE) isolates and their susceptibility to ceftazidime-avibactam and aztreonam-avibactam combinations. Nonduplicate CNSE clinical isolates were prospectively collected from six hospitals between June 2012 and November 2013. MICs of ceftazidime and aztreonam, alone or combined with a fixed concentration of avibactam (4 μg/ml), and those of carbapenems (comparator agents) were determined. MICs of ertapenem in combination with phenylalanine arginine-naphthylamide dihydrochloride (PAβN) were also determined to assess active efflux. Genes encoding carbapenemases, plasmid-mediated AmpC enzymes, extended-spectrum β-lactamases (ESBLs), and major outer membrane proteins (OMPs) were amplified and sequenced. OMPs were also extracted for SDS-PAGE analysis. Among the 139 CNSE isolates, mainly Enterobacter spp. and Klebsiella pneumoniae, 123 (88.4%) were ertapenem nonsusceptible, 12 (8.6%) exhibited reduced susceptibility to all carbapenems, and 4 Proteeae isolates (2.9%) were resistant to imipenem. Carbapenemase production was detected in only two isolates (producing OXA-48 and IMI-3). In contrast, OMP deficiency, in association with AmpCs and/or ESBLs (mainly CTX-M-9, SHV-12, and CTX-M-15), was largely identified among CNSE isolates. The ceftazidime-avibactam and aztreonam-avibactam combinations exhibited potent activity against CNSE isolates (MIC50/MIC90, 1/1 μg/ml and 0.5/0.5 μg/ml, respectively) compared to that of ceftazidime and aztreonam alone (MIC50/MIC90, 512/512 μg/ml and 128/512 μg/ml, respectively). This study reveals the in vitro activity of ceftazidime-avibactam and aztreonam-avibactam combinations against a large collection of porin-deficient enterobacterial isolates that are representative of the CNSE recovered in the northern part of France.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aztreonam, ≥98% (HPLC)