Merck
  • Evaluation of angiopoietin 1 and 2, vascular endothelial growth factor, and tumor necrosis factor alpha levels in asthmatic children.
Evaluation of angiopoietin 1 and 2, vascular endothelial growth factor, and tumor necrosis factor alpha levels in asthmatic children.
Allergy and asthma proceedings (2015-01-15)
Burcu Tahire Köksal, Ozlem Yilmaz Ozbek, Nilufer Bayraktar, Ayse Canan Yazici
RESUMO
Asthma is characterized by chronic airway inflammation that is associated with structural changes termed airway remodeling. Recently, cytokines/mediators that augment inflammation have been attracting attention in this field. The aim of this study was to evaluate serum angiopoietin (Ang)-1, Ang-2, vascular endothelial growth factor (VEGF), and tumor necrosis factor (TNF) alpha values, which have important roles in inflammation, angiogenesis, and remodeling in asthmatic children. We also documented correlations between demographic features, duration of asthma, and pulmonary function test (PFT) parameters. Randomly selected 40 children (20 male and 20 female children, aged 6-16 years) with mild or moderate persistent asthma and 32 healthy children (15 male and 17 female children, aged 6-16 years) enrolled in the study. All asthmatic children had been using inhaled corticosteroids at least for the last 3 months. Serum Ang-1 levels were significantly lower in asthmatic children than those in normal controls. The Ang-1/Ang-2 ratio was also significantly lower in asthmatic children compared with those in normal controls (p < 0.01). However, serum Ang-2, VEGF, and TNF-alpha levels were similar in the two groups. A significant positive correlation was found between VEGF and duration of asthma. No correlation between serum Ang-1, Ang-2, VEGF values, and PFT parameters was obtained. On the other hand, significant negative correlation was detected between serum TNF-alpha and forced expiratory volume in 1 second. We have shown that serum Ang-1 levels and Ang-1/Ang-2 ratio were significantly reduced and balance was toward Ang-2 in asthmatics children. This process may lead to inflammation, destabilization of blood vessels, and trigger remodeling.